Learn More
6,9-Diamino-2-ethoxyacridine lactate monohydrate, 95%, Thermo Scientific Chemicals
$56.78 - $91.05
Chemical Identifiers
| CAS | 6402-23-9 |
|---|---|
| Molecular Formula | C3H6O3·H2O |
| Molecular Weight (g/mol) | 361.39 |
| MDL Number | MFCD00149646 |
| InChI Key | NYEPHMYJRNWPLA-UHFFFAOYSA-N |
| Synonym | ethacridine lactate monohydrate, ethodin, ethacridine lactate hydrate, ethacridine lactate salt, dsstox_cid_26274, dsstox_rid_81501, dsstox_gsid_46274, 7-ethoxyacridine-3,9-diamine; 2-hydroxypropanoic acid; hydrate, rivanol monohydrate |
| PubChem CID | 165457 |
| IUPAC Name | 7-ethoxyacridine-3,9-diamine;2-hydroxypropanoic acid;hydrate |
| SMILES | CCOC1=CC2=C(C3=C(C=C(C=C3)N)N=C2C=C1)N.CC(C(=O)O)O.O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|
AC406380050
|
Thermo Scientific Chemicals
406380050 |
5 g | Glass bottle |
Each for $56.78
|
|
||||
|
AC406380100
|
Thermo Scientific Chemicals
406380100 |
10 g | Glass bottle |
Each for $91.05
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 6402-23-9 | |
| 361.39 | |
| NYEPHMYJRNWPLA-UHFFFAOYSA-N | |
| 165457 | |
| CCOC1=CC2=C(C3=C(C=C(C=C3)N)N=C2C=C1)N.CC(C(=O)O)O.O |
| C3H6O3·H2O | |
| MFCD00149646 | |
| ethacridine lactate monohydrate, ethodin, ethacridine lactate hydrate, ethacridine lactate salt, dsstox_cid_26274, dsstox_rid_81501, dsstox_gsid_46274, 7-ethoxyacridine-3,9-diamine; 2-hydroxypropanoic acid; hydrate, rivanol monohydrate | |
| 7-ethoxyacridine-3,9-diamine;2-hydroxypropanoic acid;hydrate |
Specifications
| 6402-23-9,1837-57-6 | |
| 100.0 | |
| Yellow | |
| 95% | |
| C3H6O3·H2O | |
| 5 g | |
| ethacridine lactate monohydrate, ethodin, ethacridine lactate hydrate, ethacridine lactate salt, dsstox_cid_26274, dsstox_rid_81501, dsstox_gsid_46274, 7-ethoxyacridine-3,9-diamine; 2-hydroxypropanoic acid; hydrate, rivanol monohydrate | |
| NYEPHMYJRNWPLA-UHFFFAOYSA-N | |
| 7-ethoxyacridine-3,9-diamine;2-hydroxypropanoic acid;hydrate | |
| 165457 | |
| 95% | |
| 6, 9-Diamino-2-ethoxyacridine lactate monohydrate |
| 94.0 | |
| 243°C to 245°C | |
| Authentic | |
| Glass bottle | |
| MFCD00149646 | |
| 15, 3771 | |
| Solubility in water: .. Other solubilities: slowly soluble in 15 parts water,soluble in 9 parts boiling water,soluble in 110 parts alcohol (22°C),soluble in 100 parts alcohol (20°C) | |
| CCOC1=CC2=C(C3=C(C=C(C=C3)N)N=C2C=C1)N.CC(C(=O)O)O.O | |
| 361.39 | |
| 361.39 | |
| Powder |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only