missing translation for 'onlineSavingsMsg'
Learn More

6,8-Dimethoxy-4-methylquinoline Hydrate 98.0+%, TCI America™

Catalog No. D21905G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21905G 5 g
D21901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21905G Supplier TCI America Supplier No. D21905G
Only null left

Chemical Identifiers

CAS 51049-14-0
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD00191629
InChI Key DZXKOXPAULZIGL-UHFFFAOYSA-N
PubChem CID 44630253
IUPAC Name 6,8-dimethoxy-4-methylquinoline
SMILES CC1=C2C=C(C=C(C2=NC=C1)OC)OC

Specifications

Melting Point 65°C
Quantity 5 g
Formula Weight 203.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6,8-Dimethoxy-4-methylquinoline Hydrate