accessibility menu, dialog, popup

UserName

CH3NOH3COCH3

CAS RN 51049-14-0

IUPAC Name: 6,8-dimethoxy-4-methylquinoline
Synonyms: 6,8-dimethoxy-4-methylquinoline
Molecular Weight (g/mol): 203.24
Molecular Formula: C12H13NO2
InChi Key: DZXKOXPAULZIGL-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C2N=CC=C(C)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results