missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6,8-Dibromoimidazo[1,2-a]pyrazine 98.0+%, TCI America™
Supplier: TCI America D49675G
Specifications
| 6,8-Dibromoimidazo[1,2-a]pyrazine | |
| 167°C | |
| C6H3Br2N3 | |
| 5 g | |
| UQCZZGIPIMJBCL-UHFFFAOYSA-N | |
| 6,8-dibromoimidazo[1,2-a]pyrazine | |
| 15025843 | |
| ≥98.0% (GC) |
| 63744-22-9 | |
| Yellow | |
| MFCD08460056 | |
| 6,8-dibromoimidazo 1,2-a pyrazine, imidazo 1,2-a pyrazine, 6,8-dibromo, 6,8-dibromo-imidazo 1,2-a pyrazine, 6,8-dibromoimidazol 1,2-a pyrazine, pubchem14656, acmc-1b8pv, ksc351o9d, buttpark 180\02-82, 6,8 dibromoimidazo 1,2-a pyrazine, 6,8-dibomoimidazo 1,2-a pyrazine | |
| BrC1=CN2C=CN=C2C(Br)=N1 | |
| 276.92 | |
| 276.92 | |
| Crystalline Powder |
Chemical Identifiers
| 63744-22-9 | |
| 276.92 | |
| UQCZZGIPIMJBCL-UHFFFAOYSA-N | |
| 15025843 | |
| BrC1=CN2C=CN=C2C(Br)=N1 |
| C6H3Br2N3 | |
| MFCD08460056 | |
| 6,8-dibromoimidazo 1,2-a pyrazine, imidazo 1,2-a pyrazine, 6,8-dibromo, 6,8-dibromo-imidazo 1,2-a pyrazine, 6,8-dibromoimidazol 1,2-a pyrazine, pubchem14656, acmc-1b8pv, ksc351o9d, buttpark 180\02-82, 6,8 dibromoimidazo 1,2-a pyrazine, 6,8-dibomoimidazo 1,2-a pyrazine | |
| 6,8-dibromoimidazo[1,2-a]pyrazine |
Safety and Handling
TSCA : No