missing translation for 'onlineSavingsMsg'
Learn More

6,8-Dibromoimidazo[1,2-a]pyrazine 98.0+%, TCI America™

Catalog No. p-7128578
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49671G 1 g
D49675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49671G Supplier TCI America Supplier No. D49671G
Only null left

Chemical Identifiers

CAS 63744-22-9
Molecular Formula C6H3Br2N3
Molecular Weight (g/mol) 276.92
MDL Number MFCD08460056
InChI Key UQCZZGIPIMJBCL-UHFFFAOYSA-N
Synonym 6,8-dibromoimidazo 1,2-a pyrazine, imidazo 1,2-a pyrazine, 6,8-dibromo, 6,8-dibromo-imidazo 1,2-a pyrazine, 6,8-dibromoimidazol 1,2-a pyrazine, pubchem14656, acmc-1b8pv, ksc351o9d, buttpark 180\02-82, 6,8 dibromoimidazo 1,2-a pyrazine, 6,8-dibomoimidazo 1,2-a pyrazine
PubChem CID 15025843
IUPAC Name 6,8-dibromoimidazo[1,2-a]pyrazine
SMILES BrC1=CN2C=CN=C2C(Br)=N1

Specifications

Melting Point 167°C
Color Yellow
Quantity 1 g
Formula Weight 276.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6,8-Dibromoimidazo[1,2-a]pyrazine