missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6,8-Bis(benzylthio)octanoic Acid 98.0+%, TCI America™
$212.64 - $736.66
Chemical Identifiers
| CAS | 95809-78-2 |
|---|---|
| Molecular Formula | C22H28O2S2 |
| Molecular Weight (g/mol) | 388.58 |
| MDL Number | MFCD22420826 |
| InChI Key | ZYRLHJIMTROTBO-UHFFFAOYNA-N |
| Synonym | CPI 613 |
| PubChem CID | 24770514 |
| IUPAC Name | 6,8-bis(benzylsulfanyl)octanoic acid |
| SMILES | OC(=O)CCCCC(CCSCC1=CC=CC=C1)SCC1=CC=CC=C1 |
Chemical Identifiers
| 95809-78-2 | |
| 388.58 | |
| ZYRLHJIMTROTBO-UHFFFAOYNA-N | |
| 24770514 | |
| OC(=O)CCCCC(CCSCC1=CC=CC=C1)SCC1=CC=CC=C1 |
| C22H28O2S2 | |
| MFCD22420826 | |
| CPI 613 | |
| 6,8-bis(benzylsulfanyl)octanoic acid |
Specifications
| 95809-78-2 | |
| White-Yellow | |
| MFCD22420826 | |
| CPI 613 | |
| OC(=O)CCCCC(CCSCC1=CC=CC=C1)SCC1=CC=CC=C1 | |
| 388.58 | |
| 388.58 | |
| Crystalline Powder |
| 70°C | |
| C22H28O2S2 | |
| 5 mg | |
| ZYRLHJIMTROTBO-UHFFFAOYNA-N | |
| 6,8-bis(benzylsulfanyl)octanoic acid | |
| 24770514 | |
| ≥98.0% (HPLC,T) | |
| 6,8-Bis(benzylthio)octanoic Acid |
Safety and Handling
TSCA : No