missing translation for 'onlineSavingsMsg'
Learn More

6-[[7-(N,N-Dimethylaminosulfonyl)-2,1,3-benzoxadiazol-4-yl]amino]hexanoic Acid 98.0+%, TCI America™

Catalog No. D3178100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3178100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3178100MG Supplier TCI America Supplier No. D3178100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1820741-40-9
Molecular Formula C14H20N4O5S
Molecular Weight (g/mol) 356.397
InChI Key ODEWENLZBOAUCF-UHFFFAOYSA-N
Synonym 4-(5-Carboxypentylamino)-7-(N,N-dimethylaminosulfonyl)-2,1,3-benzoxadiazole
PubChem CID 44629791
IUPAC Name 6-[[4-(dimethylsulfamoyl)-2,1,3-benzoxadiazol-7-yl]amino]hexanoic acid
SMILES CN(C)S(=O)(=O)C1=CC=C(C2=NON=C12)NCCCCCC(=O)O

Specifications

Melting Point 154°C
Color Yellow
Quantity 100 mg
Formula Weight 356.40
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-[[7-(N,N-Dimethylaminosulfonyl)-2,1,3-benzoxadiazol-4-yl]amino]hexanoic Acid