accessibility menu, dialog, popup

UserName

CAS RN 1820741-40-9

IUPAC Name: 6-{[7-(dimethylsulfamoyl)-2,1,3-benzoxadiazol-4-yl]amino}hexanoic acid
Synonyms: 6-{[7-(dimethylsulfamoyl)-2,1,3-benzoxadiazol-4-yl]amino}hexanoic acid
Molecular Weight (g/mol): 356.4
Molecular Formula: C14H20N4O5S
InChi Key: ODEWENLZBOAUCF-UHFFFAOYSA-N
SMILES: CN(C)S(=O)(=O)C1=CC=C(NCCCCCC(O)=O)C2=NON=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results