missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxy-4-methylcoumarin 98.0+%, TCI America™

Catalog No. D43901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43901G 1 g
D43905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43901G Supplier TCI America Supplier No. D43901G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4281-40-7
Molecular Formula C12H12O4
Molecular Weight (g/mol) 220.224
MDL Number MFCD00012338
InChI Key GBYDSYPGGDKWGZ-UHFFFAOYSA-N
PubChem CID 77966
IUPAC Name 6,7-dimethoxy-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=CC(=C(C=C12)OC)OC

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 1 g
Formula Weight 220.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6,7-Dimethoxy-4-methylcoumarin