accessibility menu, dialog, popup

UserName

H3COOOCH3OCH3

CAS RN 4281-40-7

IUPAC Name: 6,7-dimethoxy-4-methyl-2H-chromen-2-one
Synonyms: 6,7-dimethoxy-4-methylchromen-2-one
Molecular Weight (g/mol): 220.22
Molecular Formula: C12H12O4
InChi Key: GBYDSYPGGDKWGZ-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C(C)=CC(=O)OC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results