missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline Hydrochloride 98.0+%, TCI America™
Supplier: TCI America D247225G
Specifications
| 6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline Hydrochloride | |
| 248°C | |
| MFCD00012744 | |
| 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, heliamine hydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolinehydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydro-isoquinoline hydrochloride, 1,2,3,4-tetrahydro-6,7-dimethoxy-isoquinoline hydrochloride, isoquinoline, 1,2,3,4-tetrahydro-6,7-dimethoxy-, hydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline, chloride, pubchem22149, acmc-1cjku, ksc203s7f | |
| COC1=C(C=C2CNCCC2=C1)OC.Cl | |
| 229.70 | |
| 229.70 | |
| Crystalline Powder |
| 2328-12-3 | |
| C11H16ClNO2 | |
| 25 g | |
| SHOWAGCIRTUYNA-UHFFFAOYSA-N | |
| 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride | |
| 6456035 | |
| ≥98.0% (T) |
Chemical Identifiers
| 2328-12-3 | |
| 229.70 | |
| SHOWAGCIRTUYNA-UHFFFAOYSA-N | |
| 6456035 | |
| COC1=C(C=C2CNCCC2=C1)OC.Cl |
| C11H16ClNO2 | |
| MFCD00012744 | |
| 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, heliamine hydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolinehydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydro-isoquinoline hydrochloride, 1,2,3,4-tetrahydro-6,7-dimethoxy-isoquinoline hydrochloride, isoquinoline, 1,2,3,4-tetrahydro-6,7-dimethoxy-, hydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline, chloride, pubchem22149, acmc-1cjku, ksc203s7f | |
| 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride |
Safety and Handling
RTECSNumber : NX5017980
TSCA : No