accessibility menu, dialog, popup

UserName

CAS RN 2328-12-3

IUPAC Name: 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-ium chloride
Synonyms: 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-ium chloride
Molecular Weight (g/mol): 229.7
Molecular Formula: C11H16ClNO2
InChi Key: SHOWAGCIRTUYNA-UHFFFAOYSA-N
SMILES: [Cl-].COC1=CC2=C(C[NH2+]CC2)C=C1OC
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)