missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97%

Catalog No. p-7026463
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2246906 5 g
AAB2246914 25 g
AAB2246922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2246906 Supplier Thermo Scientific Chemicals Supplier No. B2246906
Only null left

CAS: 2328-12-3 | C11H16ClNO2 | 229.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2328-12-3
Molecular Formula C11H16ClNO2
Molecular Weight (g/mol) 229.70
MDL Number MFCD00012744
InChI Key SHOWAGCIRTUYNA-UHFFFAOYSA-N

Specifications

Melting Point 256°C to 260°C
Quantity 5 g
Beilstein 3634126
Synonym 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, heliamine hydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolinehydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydro-isoquinoline hydrochloride, 1,2,3,4-tetrahydro-6,7-dimethoxy-isoquinoline hydrochloride, isoquinoline, 1,2,3,4-tetrahydro-6,7-dimethoxy-, hydrochloride, 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline, chloride, pubchem22149, acmc-1cjku, ksc203s7f
SMILES COC1=C(C=C2CNCCC2=C1)OC.Cl
IUPAC Name 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride
PubChem CID 6456035
Formula Weight 229.71
Percent Purity 97%
Chemical Name or Material 6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber NX5017980
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only