missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dihydroxycoumarin, 98+%

CAS: 305-01-1 | C9H6O4 | 178.143 g/mol

$134.33 - $524.30

Chemical Identifiers

CAS 305-01-1
Molecular Formula C9H6O4
Molecular Weight (g/mol) 178.143
MDL Number MFCD00006874
InChI Key ILEDWLMCKZNDJK-UHFFFAOYSA-N
Synonym esculetin, 6,7-dihydroxycoumarin, aesculetin, cichorigenin, 6,7-dihydroxy-2h-chromen-2-one, cichoriin aglucon, esculatin, esculetol, esculin aglucon, esculin aglycon
PubChem CID 5281416
ChEBI CHEBI:490095
IUPAC Name 6,7-dihydroxychromen-2-one
SMILES C1=CC(=O)OC2=CC(=C(C=C21)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1539303
View Documents
Thermo Scientific Chemicals
A1539303
1 g
Each for $134.33
Only null left
 
AAA1539306
View Documents
Thermo Scientific Chemicals
A1539306
5 g
Each for $524.30
Only null left
 
Description

Description

In the presence of esculetin and HA14-1 (sc-205911) expression of the death receptor 4 (DR4) has been observed to be upregulated and extracellular-regulated kinase (ERK) to be activated. Other studies suggest that esculetin upregulates death receptor 5 (DR5) protein expression, and enhances TRAIL-induced apoptosis. It also inhibits the activity of 12-LO (12-lipoxygenase), and decreases leukotriene biosynthesis during 5-LO inhibition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In the presence of esculetin and HA14-1 (sc-205911) expression of the death receptor 4 (DR4) has been observed to be upregulated and extracellular-regulated kinase (ERK) to be activated. Other studies suggest that esculetin upregulates death receptor 5 (DR5) protein expression, and enhances TRAIL-induced apoptosis. It also inhibits the activity of 12-LO (12-lipoxygenase), and decreases leukotriene biosynthesis during 5-LO inhibition.

Solubility
Soluble in DMSO, DMF, diluted alkalies with blue fluorescence, and acetone. Slightly soluble in water. Insoluble in ether. Moderately soluble in Glacial acetic acid, and hot ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications

Chemical Identifiers

305-01-1
178.143
ILEDWLMCKZNDJK-UHFFFAOYSA-N
5281416
6,7-dihydroxychromen-2-one
C9H6O4
MFCD00006874
esculetin, 6,7-dihydroxycoumarin, aesculetin, cichorigenin, 6,7-dihydroxy-2h-chromen-2-one, cichoriin aglucon, esculatin, esculetol, esculin aglucon, esculin aglycon
CHEBI:490095
C1=CC(=O)OC2=CC(=C(C=C21)O)O

Specifications

305-01-1
C9H6O4
1 g
14,3697
Soluble in DMSO,DMF,diluted alkalies with blue fluorescence,and acetone. Slightly soluble in water. Insoluble in ether. Moderately soluble in Glacial acetic acid,and hot ethanol.
C1=CC(=O)OC2=CC(=C(C=C21)O)O
178.143
CHEBI:490095
≥98%
∼270°C (decomposition)
MFCD00006874
152788
esculetin, 6,7-dihydroxycoumarin, aesculetin, cichorigenin, 6,7-dihydroxy-2h-chromen-2-one, cichoriin aglucon, esculatin, esculetol, esculin aglucon, esculin aglycon
ILEDWLMCKZNDJK-UHFFFAOYSA-N
6,7-dihydroxychromen-2-one
5281416
178.15
6,7-Dihydroxycoumarin
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 206-161-5

RTECSNumber : GN6382500

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only