missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dihydro-5H-cyclopenta[b]pyridine 98.0+%, TCI America™

Catalog No. D35365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35365G 5 g
D353625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35365G Supplier TCI America Supplier No. D35365G

Chemical Identifiers

CAS 533-37-9
Molecular Formula C8H9N
Molecular Weight (g/mol) 119.167
MDL Number MFCD00005933
InChI Key KRNSYSYRLQDHDK-UHFFFAOYSA-N
Synonym 2,3-cyclopentenopyridine, 6,7-dihydro-5h-cyclopenta b pyridine, pyrindan, 2,3-cyclopenteno pyridine, 5h-1-pyrindine, 6,7-dihydro, 5h,6h,7h-cyclopenta b pyridine, 5h-cyclopenta b pyridine, 6,7-dihydro, pubchem15900, 2,3-cyclopentapyridine
PubChem CID 68292
IUPAC Name 6,7-dihydro-5H-cyclopenta[b]pyridine
SMILES C1CC2=C(C1)N=CC=C2

Specifications

Boiling Point 88°C
Quantity 5 g
Formula Weight 119.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 6,7-Dihydro-5H-cyclopenta[b]pyridine