missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dihydro-1H-indeno[5,4-b]furan-8(2H)-one 98.0+%, TCI America™

Catalog No. p-7128641
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5069200MG 200 mg
D50691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5069200MG Supplier TCI America Supplier No. D5069200MG

Chemical Identifiers

CAS 196597-78-1
Molecular Formula C11H10O2
Molecular Weight (g/mol) 174.199
MDL Number MFCD09955085
InChI Key ZZUIZMWFNOKNLN-UHFFFAOYSA-N
Synonym 1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-one
PubChem CID 11137616
IUPAC Name 1,2,6,7-tetrahydrocyclopenta[e][1]benzofuran-8-one
SMILES C1CC(=O)C2=C1C=CC3=C2CCO3

Specifications

Melting Point 135°C
Color White-Yellow
Quantity 200 mg
Formula Weight 174.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6,7-Dihydro-1H-indeno[5,4-b]furan-8(2H)-one