accessibility menu, dialog, popup

UserName

CAS RN 196597-78-1

IUPAC Name: 1H,2H,6H,7H,8H-indeno[5,4-b]furan-8-one
Synonyms: 1H,2H,6H,7H-indeno[5,4-b]furan-8-one
Molecular Weight (g/mol): 174.2
Molecular Formula: C11H10O2
InChi Key: ZZUIZMWFNOKNLN-UHFFFAOYSA-N
SMILES: O=C1CCC2=C1C1=C(OCC1)C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results