missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6,7,8,9-Tetrahydro-5H-benzocyclohepten-5-one 97.0+%, TCI America™
Supplier: TCI America T13475G
Specifications
| 6,7,8,9-Tetrahydro-5H-benzocyclohepten-5-one | |
| Yellow | |
| C11H12O | |
| 5 g | |
| KWHUHTFXMNQHAA-UHFFFAOYSA-N | |
| 6,7,8,9-tetrahydro-5H-benzo[7]annulen-5-one | |
| 70003 | |
| ≥97.0% (GC) |
| 826-73-3 | |
| 270°C | |
| MFCD00004144 | |
| 1-Benzosuberone | |
| O=C1CCCCC2=CC=CC=C12 | |
| 160.22 | |
| 160.22 | |
| Liquid |
Chemical Identifiers
| 826-73-3 | |
| 160.22 | |
| KWHUHTFXMNQHAA-UHFFFAOYSA-N | |
| 70003 | |
| O=C1CCCCC2=CC=CC=C12 |
| C11H12O | |
| MFCD00004144 | |
| 1-Benzosuberone | |
| 6,7,8,9-tetrahydro-5H-benzo[7]annulen-5-one |
Safety and Handling
TSCA : No