missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6,7,8,9-Tetrahydro-5H-benzocyclohepten-5-one 97.0+%, TCI America™
$214.37 - $684.11
Chemical Identifiers
| CAS | 826-73-3 |
|---|---|
| Molecular Formula | C11H12O |
| Molecular Weight (g/mol) | 160.22 |
| MDL Number | MFCD00004144 |
| InChI Key | KWHUHTFXMNQHAA-UHFFFAOYSA-N |
| Synonym | 1-Benzosuberone |
| PubChem CID | 70003 |
| IUPAC Name | 6,7,8,9-tetrahydro-5H-benzo[7]annulen-5-one |
| SMILES | O=C1CCCCC2=CC=CC=C12 |
Chemical Identifiers
| 826-73-3 | |
| 160.22 | |
| KWHUHTFXMNQHAA-UHFFFAOYSA-N | |
| 70003 | |
| O=C1CCCCC2=CC=CC=C12 |
| C11H12O | |
| MFCD00004144 | |
| 1-Benzosuberone | |
| 6,7,8,9-tetrahydro-5H-benzo[7]annulen-5-one |
Specifications
| 826-73-3 | |
| 270°C | |
| MFCD00004144 | |
| 1-Benzosuberone | |
| O=C1CCCCC2=CC=CC=C12 | |
| 160.22 | |
| 160.22 | |
| Liquid |
| Yellow | |
| C11H12O | |
| 5 g | |
| KWHUHTFXMNQHAA-UHFFFAOYSA-N | |
| 6,7,8,9-tetrahydro-5H-benzo[7]annulen-5-one | |
| 70003 | |
| ≥97.0% (GC) | |
| 6,7,8,9-Tetrahydro-5H-benzocyclohepten-5-one |
Safety and Handling
TSCA : No