missing translation for 'onlineSavingsMsg'
Learn More

[6,6]-Phenyl-C71-butyric Acid Methyl Ester (mixture of isomers) 98.0+%, TCI America™

Catalog No. M255050MG
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M255050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M255050MG Supplier TCI America Supplier No. M255050MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 609771-63-3
Molecular Formula C82H14O2
Molecular Weight (g/mol) 1031.012
MDL Number MFCD09842750
InChI Key AZSFNTBGCTUQFX-UHFFFAOYSA-N
Synonym Methyl [6,6]-Phenyl-C71-butyrate, [70]PCBM
PubChem CID 71777692
SMILES COC(=O)CCCC1(C23C14C5=C6C7=C8C9=C1C%10=C%11C9=C9C%12=C%13C%14=C%15C%16=C%17C%18=C%19C%20=C%21C%22=C%23C%24=C%25C%26=C%27C%28=C(C%14=C%14C%12=C%11C%11=C%14C%28=C%26C%12=C%11C%10=C%10C%12=C%25C%23=C%11C%10=C1C7=C%11C%22=C6C4=C%21C%19=C2C%17=C1C%15=C%13C2=C9

Specifications

Color Black
Quantity 50 mg
Formula Weight 1031.01
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material [6,6]-Phenyl-C71-butyric Acid Methyl Ester (mixture of isomers)