missing translation for 'onlineSavingsMsg'
Learn More

[6,6]-Phenyl-C61-butyric Acid Dodecyl Ester 98.0+%, TCI America™

Catalog No. P2015100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2015100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2015100MG Supplier TCI America Supplier No. P2015100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 571177-69-0
Molecular Formula C83H36O2
Synonym [60]PCB-C12

Specifications

Color Black
Quantity 100 mg
Formula Weight 1065.20
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material [6,6]-Phenyl-C61-butyric Acid Dodecyl Ester