missing translation for 'onlineSavingsMsg'
Learn More

[6,6]-Phenyl-C61-butyric Acid Butyl Ester 98.0+%, TCI America™

Catalog No. P2013100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2013100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2013100MG Supplier TCI America Supplier No. P2013100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 571177-66-7
Molecular Formula C75H20O2
MDL Number MFCD09842755
Synonym PCBB, [60]PCB-C4

Specifications

Quantity 100 mg
Formula Weight 952.98
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material [6,6]-Phenyl-C61-butyric Acid Butyl Ester