missing translation for 'onlineSavingsMsg'
Learn More

6,6'-Dithiodinicotinic Acid 98.0+%, TCI America™

Catalog No. p-7127370
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25645G 5 g
D256425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25645G Supplier TCI America Supplier No. D25645G

Chemical Identifiers

CAS 15658-35-2
Molecular Formula C12H8N2O4S2
Molecular Weight (g/mol) 308.33
MDL Number MFCD00006454
InChI Key GSASOFRDSIKDSN-UHFFFAOYSA-N
PubChem CID 85040
IUPAC Name 6-[(5-carboxypyridin-2-yl)disulfanyl]pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=C(SSC2=CC=C(C=N2)C(O)=O)N=C1

Specifications

Color White-Yellow
Quantity 5 g
Synonym 6,6′C-Dithiobis(3-pyridinecarboxylic Acid)
Formula Weight 308.33
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6,6′-Dithiodinicotinic Acid
RTECSNumber US5684700
TSCA No