missing translation for 'onlineSavingsMsg'
Learn More

6,6-Dimethyl-5,7-dioxaspiro[2.5]octane-4,8-dione 98.0+%, TCI America™

Catalog No. p-7124919
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13341G 1 g
C13345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13341G Supplier TCI America Supplier No. C13341G
Only null left

Chemical Identifiers

CAS 5617-70-9
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.16
MDL Number MFCD00042796
InChI Key AXJVPXNVESYGDT-UHFFFAOYSA-N
PubChem CID 79720
IUPAC Name 6,6-dimethyl-5,7-dioxaspiro[2.5]octane-4,8-dione
SMILES CC1(C)OC(=O)C2(CC2)C(=O)O1

Specifications

Melting Point 70°C
Color White-Yellow
Quantity 1 g
Synonym 6,6-dimethyl-5,7-dioxaspiro 2.5 octane-4,8-dione, cycl-isopropylidene cyclopropane-1,1-dicarboxylate, 5,7-dioxaspiro 2.5 octane-4,8-dione, 6,6-dimethyl, 5,7-dioxaspiro 2.5 octane-4,8-dione,6,6-dimethyl, 6,6-dimethyl-5,7-dioxaspiro 2,5 octane-4,8-dione, acmc-1az6b, isopropylidene cyclopropane-1,1-dicarboxylate, cyclic isopropylidene 1,1-cyclopropanedicarboxylate
Formula Weight 170.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6,6-Dimethyl-5,7-dioxaspiro[2.5]octane-4,8-dione