accessibility menu, dialog, popup

UserName

CAS RN 5617-70-9

IUPAC Name: 6,6-dimethyl-5,7-dioxaspiro[2.5]octane-4,8-dione
Synonyms: 6,6-dimethyl-5,7-dioxaspiro[2.5]octane-4,8-dione
Molecular Weight (g/mol): 170.16
Molecular Formula: C8H10O4
InChi Key: AXJVPXNVESYGDT-UHFFFAOYSA-N
SMILES: CC1(C)OC(=O)C2(CC2)C(=O)O1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)