missing translation for 'onlineSavingsMsg'
Learn More

6,6-Dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione, 98%

Catalog No. p-7056960
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6621203 1 g
AAH6621206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6621203 Supplier Thermo Scientific Chemicals Supplier No. H6621203

CAS: 194421-56-2 | C7H9NO2 | 139.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 194421-56-2
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD12923551
InChI Key DRMNZTFLOOSXIN-UHFFFAOYSA-N
PubChem CID 10749252
IUPAC Name 6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione
SMILES CC1(C2C1C(=O)NC2=O)C

Specifications

Quantity 1 g
Beilstein 117393
Synonym 6,6-dimethyl-3-azabicyclo 3.1.0 hexane-2,4-dione, 3,3-dimethylcyclopropane-1,2-dicarboximide, 3-azabicyclo 3.1.0 hexane-2,4-dione, 6,6-dimethyl, 3-azabicyclo 3.1.0 hexane-2,4-dione,6,6-dimethyl
Formula Weight 139.15
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 6,6-Dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only