missing translation for 'onlineSavingsMsg'
Learn More

6(5H)-Phenanthridinone 98.0+%, TCI America™

Catalog No. P19981G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19981G 1 g
P19985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P19981G Supplier TCI America Supplier No. P19981G

Chemical Identifiers

CAS 1015-89-0
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.221
MDL Number MFCD00004988
InChI Key RZFVLEJOHSLEFR-UHFFFAOYSA-N
Synonym 6 5h-phenanthridinone, phenanthridin-6 5h-one, 6-phenanthridinol, phenanthridone, 6-5h-phenanthridinone, 6-phenanthridone, 6-phenanthridinone, 6 5h-phenanthridone, phenanthridin-6-ol, phenantridone
PubChem CID 1853
ChEBI CHEBI:75292
IUPAC Name 5H-phenanthridin-6-one
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3NC2=O

Specifications

Melting Point 295°C
Quantity 1 g
Formula Weight 195.22
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 6(5H)-Phenanthridinone
RTECSNumber SG0370000
TSCA No