accessibility menu, dialog, popup

UserName

CAS RN 1015-89-0

IUPAC Name: 5,6-dihydrophenanthridin-6-one
Synonyms: 5H-phenanthridin-6-one
Molecular Weight (g/mol): 195.22
Molecular Formula: C13H9NO
InChi Key: RZFVLEJOHSLEFR-UHFFFAOYSA-N
SMILES: O=C1NC2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)