missing translation for 'onlineSavingsMsg'
Learn More

6,13-Pentacenedione 95.0+%, TCI America™

Catalog No. p-7135207
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16855G 5 g
P168525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16855G Supplier TCI America Supplier No. P16855G
Only null left

Chemical Identifiers

CAS 3029-32-1
Molecular Formula C22H12O2
Molecular Weight (g/mol) 308.336
MDL Number MFCD00003709
InChI Key UFCVADNIXDUEFZ-UHFFFAOYSA-N
Synonym 6,13-pentacenequinone, 6,13-pentacenedione, acmc-209hec, ufcvadnixduefz-uhfffaoysa
PubChem CID 76415
IUPAC Name pentacene-6,13-dione
SMILES C1=CC=C2C=C3C(=CC2=C1)C(=O)C4=CC5=CC=CC=C5C=C4C3=O

Specifications

Melting Point 394°C
Quantity 5 g
Formula Weight 308.34
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6,13-Pentacenedione