missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6,12-Dibromochrysene 98.0+%, TCI America™
Supplier: TCI America D42361G
Specifications
| 6,12-Dibromochrysene | |
| 284°C | |
| C18H10Br2 | |
| 1 g | |
| RULVBMDEPWAFIN-UHFFFAOYSA-N | |
| 6,12-dibromochrysene | |
| 3286225 | |
| ≥98.0% (HPLC) |
| 131222-99-6 | |
| Yellow | |
| MFCD00092281 | |
| 6,12-dibromochryscene, chrysene, 6,12-ibromo, chrysene, 6,12-dibromo, acmc-1c5x4, 6,12-bis bromanyl chrysene, 6,12-dibromochrysen | |
| BrC1=CC2=C(C=C(Br)C3=CC=CC=C23)C2=CC=CC=C12 | |
| 386.09 | |
| 386.09 | |
| Crystalline Powder |
Chemical Identifiers
| 131222-99-6 | |
| 386.09 | |
| RULVBMDEPWAFIN-UHFFFAOYSA-N | |
| 3286225 | |
| BrC1=CC2=C(C=C(Br)C3=CC=CC=C23)C2=CC=CC=C12 |
| C18H10Br2 | |
| MFCD00092281 | |
| 6,12-dibromochryscene, chrysene, 6,12-ibromo, chrysene, 6,12-dibromo, acmc-1c5x4, 6,12-bis bromanyl chrysene, 6,12-dibromochrysen | |
| 6,12-dibromochrysene |
Safety and Handling
TSCA : No