missing translation for 'onlineSavingsMsg'
Learn More

6,12-Dibromochrysene 98.0+%, TCI America™

Catalog No. D4236200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4236200MG 200 mg
D42361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4236200MG Supplier TCI America Supplier No. D4236200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 131222-99-6
Molecular Formula C18H10Br2
Molecular Weight (g/mol) 386.09
MDL Number MFCD00092281
InChI Key RULVBMDEPWAFIN-UHFFFAOYSA-N
Synonym 6,12-dibromochryscene, chrysene, 6,12-ibromo, chrysene, 6,12-dibromo, acmc-1c5x4, 6,12-bis bromanyl chrysene, 6,12-dibromochrysen
PubChem CID 3286225
IUPAC Name 6,12-dibromochrysene
SMILES BrC1=CC2=C(C=C(Br)C3=CC=CC=C23)C2=CC=CC=C12

Specifications

Melting Point 284°C
Color Yellow
Quantity 200 mg
Formula Weight 386.09
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6,12-Dibromochrysene