missing translation for 'onlineSavingsMsg'
Learn More

5X Kinase Buffer A

Catalog No. PV3189
Encompass
Change view
Click to view available options
Quantity:
4 mL
100 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3189 4 mL
PV6135 100 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PV3189 Supplier Thermo Fisher Scientific Supplier No. PV3189

5X Kinase Buffer A

The 5X Kinase Buffer A is a concentrated general kinase assay buffer that can be used with a variety of assay technologies such as, LanthaScreen™, Adapta™, and Z’-LYTE™ assays.

Ready to use and validated with LanthaScreen™, Adapta™, and Z’LYTE™ assays

  • Optimized 5X concentrated formulation
  • Simplified handling with room temperature storage

    Easily Screen for Kinase Targets using an Invitrogen™ Platform
    For more information on the kinase assays that can be used with our 5X Kinase Buffer A see the following links:
    LanthaScreen™ Eu Binding Assay
    LanthaScreen™ Activity Assay
    Adapta™ Universal Kinase Assay
    Z´-LYTE™ Kinase Assay Kits

    Save Freezer Space
    The 5X Kinase Buffer can be stored at room temperature, saving you time and precious freezer space.

    For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
  • Specifications

    Chemical Name or Material Kinase Buffer A
    Concentration 5X
    For Use With (Application) Pharma & Biopharma; Protein Assays and Analysis
    Physical Form Liquid
    Quantity 4 mL
    Includes 5X Kinase Buffer A may be shipped frozen or at room temperature. It should be stored at room temperature (20-30°C).

    For Research Use Only. Not for use in diagnostic procedures.