missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 5mL Polystyrene Immuno-Tubes
GREENER_CHOICE

Use for determining hormone levels.

$680.46

Specifications

Material Polystyrene
Product Type Immuno Tube
For Use With (Application) Determining hormone levels
Length (Metric) 75 mm
Diameter (Metric) Outer 12 mm
View More Specs

Products 1
Catalog Number Mfr. No. Surface Includes Price Quantity & Availability  
Catalog Number Mfr. No. Surface Includes Price Quantity & Availability  
07000424
GREENER_CHOICE
View Documents Promotion Details
Greiner Bio-One
115071
High-binding Star Bottom
Case of 2000 Each for $680.46
Only null left
 
Description

Description

  • Round bottom
  • Dia. x Height: 12 x 75mm
  • Available with either medium or high binding capacities
  • With or without star at bottom
  • Star at the bottom of the tubes serve to increase the surface and thus make it possible to bind amounts of antigens or antibodies
Specifications

Specifications

Polystyrene
Determining hormone levels
12 mm
Immuno Tube
75 mm
5 mL
Videos
SDS
Documents

Documents