missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 5mL Polystyrene Immuno-Tubes
GREENER_CHOICE

Catalog No. 07000424
Change view
Click to view available options
Surface:
High-binding
Includes:
Star Bottom
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Includes
07000424 High-binding Star Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07000424 Supplier Greiner Bio-One Supplier No. 115071

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use for determining hormone levels.

  • Round bottom
  • Dia. x Height: 12 x 75mm
  • Available with either medium or high binding capacities
  • With or without star at bottom
  • Star at the bottom of the tubes serve to increase the surface and thus make it possible to bind amounts of antigens or antibodies

Specifications

For Use With (Application) Determining hormone levels
Product Type Immuno Tube
Material Polystyrene
Surface High-binding
Length (Metric) 75 mm
Diameter (Metric) Outer 12 mm
Volume (Metric) 5 mL
Includes Star Bottom