missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Phthalate, NF, 98-102%, Spectrum™ Chemical

Catalog No. 18602193
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602193 200 L Amber Glass Bottle
18602190 500 mL Amber Glass Bottle
18602191 4 L Amber Glass Bottle
18602192 20 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602193 Supplier Spectrum Chemical Mfg Cor Supplier No. D1097200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

D1097, 84-66-2, C12H14O4

Spectrum™ Chemical Diethyl Phthalate, NF - Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 84-66-2
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
InChI Key FLKPEMZONWLCSK-UHFFFAOYSA-N
IUPAC Name 1,2-diethyl benzene-1,2-dicarboxylate
SMILES CCOC(=O)C1=CC=CC=C1C(=O)OCC

Specifications

CAS Max % 100%
Ignition Residue 0.02%
Packaging Amber Glass Bottle
Refractive Index 1.500-1.505
Quantity 200 L
Specific Gravity 1.118-1.122
Percent Purity 98 to 102%
Grade NF
Physical Form Liquid