missing translation for 'onlineSavingsMsg'
Learn More

5H-5-Methyl-6,7-dihydrocyclopentapyrazine 97.0+%, TCI America™

Catalog No. M10865ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10865ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M10865ML Supplier TCI America Supplier No. M10865ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23747-48-0
Molecular Formula C8H13N2
Molecular Weight (g/mol) 137.21
MDL Number MFCD00040997
InChI Key COSMFKAROKURLX-ZCFIWIBFSA-O
Synonym 5-methyl-6,7-dihydro-5h-cyclopenta b pyrazine, 5h-5-methyl-6,7-dihydrocyclopentapyrazine, 5h-cyclopentapyrazine, 6,7-dihydro-5-methyl, 6,7-dihydro-5-methyl-5h-cyclopentapyrazine, 6,7-dihydro-5-methyl-5 h-cyclopentapyrazine, fema no. 3306, 5-methyl-6,7-dihydro-5h-cyclopentapyrazine, 5h-5-methyl-6,7-dihydrocyclopenta b pyrazine
PubChem CID 32065
IUPAC Name (5R)-5-methyl-1H,2H,5H,6H,7H-cyclopenta[b]pyrazin-1-ium
SMILES C[C@@H]1CCC2=C1N=CC[NH2+]2

Specifications

Color Yellow
Boiling Point 80°C
Quantity 5 mL
Formula Weight 134.18
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5H-5-Methyl-6,7-dihydrocyclopentapyrazine