accessibility menu, dialog, popup

UserName

H3C(R)NNH2

CAS RN 23747-48-0

IUPAC Name: (5R)-5-methyl-1H,2H,5H,6H,7H-cyclopenta[b]pyrazin-1-ium
Synonyms: (5R)-5-methyl-1H,2H,5H,6H,7H-cyclopenta[b]pyrazin-1-ium
Molecular Weight (g/mol): 137.21
Molecular Formula: C8H13N2
InChi Key: COSMFKAROKURLX-ZCFIWIBFSA-O
SMILES: C[C@@H]1CCC2=C1N=CC[NH2+]2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results