missing translation for 'onlineSavingsMsg'
Learn More

5alpha-Cholestan-3-one, 97%

Catalog No. AAL0872603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0872603 1 g
AAL0872606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0872603 Supplier Thermo Scientific Chemicals Supplier No. L0872603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 566-88-1 | C27H46O | 386.664 g/mol

It is employed as a intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 566-88-1
Molecular Formula C27H46O
Molecular Weight (g/mol) 386.664
MDL Number MFCD00065901
InChI Key PESKGJQREUXSRR-UXIWKSIVSA-N
Synonym 5alpha-cholestan-3-one, 5a-cholestan-3-one, 5alpha-cholestanone, coprostanone, 5alpha-cholestane-3-one, cholestan-3-one, 5a, 3-keto-5alpha-cholestane, 5-alpha-cholestan-3-one, cholestan-3-one
PubChem CID 92128
ChEBI CHEBI:17762
IUPAC Name (5S,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-one
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4C3(CCC(=O)C4)C)C

Specifications

Melting Point 128°C to 132°C
Quantity 1 g
Beilstein 2625580
Solubility Information Insoluble in water.
Optical Rotation +43° (c=1 in Chloroform)
Formula Weight 386.66
Percent Purity 97%
Chemical Name or Material 5alpha-Cholestan-3-one
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only