accessibility menu, dialog, popup

UserName

CAS RN 566-88-1

IUPAC Name: (1R,3aS,3bR,5aS,9aS,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-hexadecahydro-1H-cyclopenta[a]phenanthren-7-one
Synonyms: coprostanone 5α-cholestanone (5α)-cholestanone 5a-cholestanone 5a(H)-cholestan-3-one 5a-cholestan-3-one
Molecular Weight (g/mol): 386.66
Molecular Formula: C27H46O
InChi Key: PESKGJQREUXSRR-UXIWKSIVSA-N
SMILES: CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results