missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 2 12-14 kD MWCO Standard RC Dry Dialysis Kits
Description
Includes
- 1m or 5m of Spectra/Por 2 dialysis tubing
- 1 Standard Spectra/Por Closure (for buoyance)
- 1 weighted Spectra/Por Closure (for orientation)
- 2 adhesive tubing openers
- For higher permeability applications, or evaluating dialysis tubing for dialysis application
- Packaged dry with glycerin (soak in water)
Specifications
Specifications
| Product Type | Trial Kit |
| MWCO | 12 to 14 kDa |
| Material | Regenerated Cellulose |
| Format | Tubing |
| Diameter (Metric) | 29 mm |
| Diameter (English) | 1.1 in. |
| Length (English) | 16 ft. |
| Length (Metric) | 5 m |
| Capacity (Metric) | 6.4 mL |
| Kit Contents | 5m roll of Spectra/Por™ 1, 2 or 3 dialysis tubing, 1 standard spectra/por closure (aids buoyancy), 1 weighted spectra/por closure (aids vertical orientation), 5 opening picks |
| Show More |