missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 1 6-8 kD MWCO Standard RC Dry Dialysis Kits

Catalog No. 08801242
Change view
Click to view available options
Diameter (Metric):
6.4 mm
14.6 mm
20.4 mm
25.5 mm
32 mm
64 mm
76 mm
Length (Metric):
0.32 mL/cm vol./length
1.7 mL/cm
32 mL/cm vol./length
46 mL/cm vol./length
5.1 mL/cm vol./length
5 m
Quantity:
5 m Tubing/Kit
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric) Quantity
08801242 32 mm 5 m Each
08700148 76 mm 46 mL/cm vol./length 5 m Tubing/Kit
08700146 64 mm 32 mL/cm vol./length 5 m Tubing/Kit
08700143 25.5 mm 5.1 mL/cm vol./length 5 m Tubing/Kit
08700140 14.6 mm 1.7 mL/cm 5 m Tubing/Kit
08700138 6.4 mm 0.32 mL/cm vol./length 5 m Tubing/Kit
08801241 20.4 mm 5 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 08801242 Supplier Spectrum™ Supplier No. 132665T

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • 1m or 5m of Spectra/Por 1 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers

With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated

  • For basic dialysis applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water)

Specifications

Product Type Trial Kit
MWCO 6 to 8 kDa
Membrane Regenerated Cellulose
Format Tubing
Diameter (Metric) 32 mm
Diameter (English) 1.2 in.
Length (English) 16 ft.
Length (Metric) 5 m
Capacity (Metric) 7.9 mL
Kit Contents 5m roll of Spectra/Por™ 1, 2 or 3 dialysis tubing, 1 standard spectra/por closure (aids buoyancy), 1 weighted spectra/por closure (aids vertical orientation), 5 opening picks
Quantity Each
For Use With (Application) Ideal for small volume dialysis or evaluation of membrane for an application
Show More Show Less