Learn More
Spectrum™ Labs Spectra/Por™ 1 6-8 kD MWCO Standard RC Dry Dialysis Kits
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
Supplier:  Spectrum™ 132660T
 View more versions of this product
Catalog No. 08-700-143
Only null left
Explore available promotions
Includes:
  • 1m or 5m of Spectra/Por 1 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers
Description
Description
  • For basic dialysis applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water)
Specifications
Specifications
6 to 8 kDa
25.5 mm
5.1 mL/cm vol./length
5 m Tubing/Kit
Product Suggestions
Product Suggestions