missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs 100 kD Biotech CE Dialysis Kits

Catalog No. 08801253
Change view
Click to view available options
Diameter (Metric):
6.4 mm
10 mm
15 mm
20 mm
Length (Metric):
0.32 mL/cm vol./length
0.79 mL/cm vol./length
1.8 mL/cm vol./length
3.1 mL/cm vol./length
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric)
08801253 10 mm 0.79 mL/cm vol./length
08700132 20 mm 3.1 mL/cm vol./length
08700131 15 mm 1.8 mL/cm vol./length
08700130 6.4 mm 0.32 mL/cm vol./length
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 08801253 Supplier Spectrum™ Supplier No. 131414T

Includes

  • 1m Biotech CE Tubing in 0.05% sodium azide
  • Two universal tubing closures
  • Five opening picks

With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane

Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC

  • Hydrophilic, symmetric porositiy
  • For small tubing demand applications or evaluating a specific dialysis membrane
  • Includes 1m tubing with pair of tubing closures and 5 opening picks

Specifications

Product Type Dialysis Trial Kit
MWCO 100 kDa
Material Cellulose Ester
Diameter (Metric) 10 mm
Diameter (English) 0.39 in.
Length (Metric) 0.79 mL/cm vol./length
Kit Contents 1m roll Biotech grade CE dialysis membrane tubing, 2 universal closures, 5 opening picks
Quantity Each
For Use With (Application) Ideal for small, critical dialysis applications