missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs 0.5 - 1.0 kD Biotech CE Dialysis Kits
Description
Includes
- 1m Biotech CE Tubing in 0.05% sodium azide
- Two universal tubing closures
- Five opening picks
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
- Hydrophilic, symmetric porositiy
- For small tubing demand applications or evaluating a specific dialysis membrane
- Includes 1m tubing with pair of tubing closures and 5 opening picks
Specifications
Specifications
| MWCO | 0.5 to 1.0 kDa |
| Diameter (Metric) | 20 mm |
| Length (Metric) | 3.1 mL/cm vol./length |
| Quantity | 1 m Tubing/Kit |