missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 7 10,000 D MWCO Standard RC Pre-Treated Dialysis Kits

With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds

$502.21 - $502.21

Specifications

MWCO 10 kDa
Quantity 1 m Tubing/Kit
View More Specs

Includes:
  • 1meter of pre-treated tubing, a pair of closures and 5 opening picks

Products 2
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Length (Metric) Price Quantity  
08-700-214
View Documents
Spectrum™
132120T
29 mm 6.4 mL/cm vol./length
Each for $502.21
Only null left
 
08-700-213
View Documents
Spectrum™
132119T
20.4 mm 3.3 mL/cm vol./length
Each for $502.21
Only null left
 
Description

Description

  • Hydrophilic, symmetric porositiy
  • Equilibrated in water
  • Packaged wet in preservative solution of 0.05% sodium azide
  • For small experiments or qualifying a membrane for a particular application

Specifications

Specifications

10 kDa
1 m Tubing/Kit
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'