Learn More
Invitrogen™ iBind™ Flex Western Device DFS Item
Allows better antigen-antibody interaction and solutions flow between the iBind™ Card and membrane for maximum interaction between protein and antibody
Supplier:  Invitrogen™ SLF2000
 View more versions of this product
Catalog No. SLF2000
Only null left
Explore available promotions
Description
Description
The iBind™ Flex Western System offers automated western blot processing and can accommodate various blot formats including midi, mini, and vertically cut strips. Based on Sequential Lateral Flow (SLF) technology, all blocking, antibody incubation and washing steps during immunodetection are hands-free. Compared to manual methods, the iBind Flex Western System requires less primary antibody, enables reproducible results and is compatible with downstream detection protocols.
The Invitrogen™ iBind™ Flex Western System offers:
  • Flexible options—optimize antibody use, change formats and reduce hands-on time
  • Cost savings—use up to 80% less primary antibody
  • Reproducibility—automated processing provides improved blot-to-blot consistency
Specifications
Specifications
1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Western Blot
iBind Flex
1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Roller
Room Temperature
Fluorescence, Chemiluminescence, Chromogenic
Nitrocellulose, PVDF
∼2.5 hr.
Western Device
1 device
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.