missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBind™ Flex Western Device

Allows better antigen-antibody interaction and solutions flow between the iBind™ Card and membrane for maximum interaction between protein and antibody

$4,675.00 - $7,660.00

Specifications

Capacity 1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Detection Method Fluorescence, Chemiluminescence, Chromogenic
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Product Line iBind Flex
View More Specs

Products 2
Catalog Number Mfr. No. Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Type Quantity Price Quantity & Availability  
SLF2000
Encompass
View Documents Promotion Details
Invitrogen™
SLF2000
Western Device 1 device
Each for $4,675.00
Only null left
 
SLF20002PK
Encompass
View Documents Promotion Details
Invitrogen™
SLF20002PK
Western Device 2 devices
Pack of 2 Each for $7,660.00
Only null left
 
Description

Description

The iBind™ Flex Western System offers automated western blot processing and can accommodate various blot formats including midi, mini, and vertically cut strips. Based on Sequential Lateral Flow (SLF) technology, all blocking, antibody incubation and washing steps during immunodetection are hands-free. Compared to manual methods, the iBind Flex Western System requires less primary antibody, enables reproducible results and is compatible with downstream detection protocols.

The Invitrogen™ iBind™ Flex Western System offers:
  • Flexible options—optimize antibody use, change formats and reduce hands-on time
  • Cost savings—use up to 80% less primary antibody
  • Reproducibility—automated processing provides improved blot-to-blot consistency
Specifications

Specifications

1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Western Blot
iBind Flex
1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Room Temperature
Fluorescence, Chemiluminescence, Chromogenic
Nitrocellulose, PVDF
∼2.5 hr.
Western Device
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.