missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 7 3500 D MWCO Standard RC Pre-Treated Dialysis Kits
Description
Includes
- 1meter of pre-treated tubing, a pair of closures and 5 opening picks
- Hydrophilic, symmetric porositiy
- Equilibrated in water
- Packaged wet in preservative solution of 0.05% sodium azide
- For small experiments or qualifying a membrane for a particular application
Specifications
Specifications
| Product Type | Dialysis Tubing Kit |
| MWCO | 3.5 kDa |
| Format | Tubing |
| Diameter (Metric) | 34 mm |
| Length (Metric) | 9.3 mL/cm vol./length |
| Quantity | 1 m Tubing/Kit |