Learn More
Agilent BioTek 50 TS Microplate Washer, IVD
The BioTek 50 TS washer offers excellent microplate washing performance. For In Vitro Diagnostic Use.
Supplier: Agilent BioTek 50TS8MVSI
Description
The BioTek 50 TS washer offers excellent microplate washing performance. Ideal for use as an ELISA washer, the system is also well suited for cell-based assay washing. The 50 TS can process biomagnetic or polystyrene bead and vacuum filtration protocols as well
The color touch screen provides a visual interface that makes creating protocols fast and intuitive. When used with the 800 TS absorbance reader or other detection system, the 50 TS delivers convenience and high-quality results to your laboratory plate washing workflows.
Specifications
| CE and TUV marked. RoHS compliant. | |
| Optional (2 to 4 L) | |
| Programmable in minutes and seconds, up to 30 min. | |
| 9.8 kg | |
| 25 to 3000 μL/well | |
| 150 to 1000 μL/well/sec. (manifold dependent) | |
| 50TS8MV | |
| 4.3 in. Color LCD Touch Screen Display |
| USB (1) | |
| 24-, 96-, 384-well Plates (96- and 384-well: Standard and Low Profile Plates, Solid and Filter Bottom (Option), 24-well Plates with 4-well Manifold) | |
| Microplate Washer | |
| 1 to 10 | |
| 4.3 in. LCD Color Touch Screen | |
| One positive displacement syringe drive | |
| 96-well plates only; Buffer Switching; Biomagnetic Separation (with available accessory module sold separately) | |
| 22 lb. |