missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek 50 TS Microplate Washer
GREENER_CHOICE

The BioTek 50 TS washer offers excellent microplate washing performance. For Research Use Only. Not for use in diagnostic procedures.

Supplier:  Agilent BioTek 50TS12VSN

 View more versions of this product

Catalog No. BT50TS12V

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

The BioTek 50 TS washer offers excellent microplate washing performance. Ideal for use as an ELISA washer, the system is also well suited for cell-based assay washing. The 50 TS can process biomagnetic or polystyrene bead and vacuum filtration protocols as well

The color touch screen provides a visual interface that makes creating protocols fast and intuitive. When used with the 800 TS absorbance reader or other detection system, the 50 TS delivers convenience and high-quality results to your laboratory plate washing workflows.

Warranty

12 months

Specifications

Chemical Identifiers

CE and TUV marked. RoHS compliant.
Optional (2 to 4 L)
Programmable in minutes and seconds, up to 30 min.
9.8 kg
25 to 3000 μL/well
150 to 1000 μL/well/sec. (manifold dependent)
4.3 in. Color LCD Touch Screen Display
USB (1)
24-, 96-, 384-well Plates (96- and 384-well: Standard and Low Profile Plates, Solid and Filter Bottom (Option), 24-well Plates with 4-well Manifold)
Microplate Washer
1 to 10
4.3 in. LCD Color Touch Screen
One positive displacement syringe drive
22 lb.

Specifications

CE and TUV marked. RoHS compliant.
Optional (2 to 4 L)
Programmable in minutes and seconds, up to 30 min.
9.8 kg
25 to 3000 μL/well
150 to 1000 μL/well/sec. (manifold dependent)
50TS12V
4.3 in. Color LCD Touch Screen Display
USB (1)
24-, 96-, 384-well Plates (96- and 384-well: Standard and Low Profile Plates, Solid and Filter Bottom (Option), 24-well Plates with 4-well Manifold)
Microplate Washer
1 to 10
4.3 in. LCD Color Touch Screen
One positive displacement syringe drive
96-well plates only; Buffer Switching
22 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions